Läs mer
Succinic Acid-1,4-13C2, TRC
Produktkod. 30425484
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425484 50 mg
30415584 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30425484 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS68876850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 79864-95-2
Molekylformel 13C2C2H6O4
Molekylvikt (g/mol) 120.07
Synonym Butanedioic Acid-13C2, 1,2-Ethanedicarboxylic Acid-13C2, 1,4-Butanedioic Acid-13C2, A 12084-13C2, Amber Acid-13C2, Asuccin-13C2, Dihydrofumaric Acid-13C2, Katasuccin-13C2, NSC 106449-13C2, NSC 25949-13C2, Wormwood Acid-13C2, Yantar-antitox-13C2
LEDER C(C[13C](=O)O)[13C](=O)O
Specifikationer
Kemiskt namn eller material Succinic Acid-1,4-13C2
Kvantitet 50 mg
InChI formel InChI=1S/C4H6O4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)/i3+1,4+1
Formel vikt 120.0333