Learn More
Succinylcholine chloride, 96%, Thermo Scientific Chemicals
Catalog Number p-7077830
Click to view available options
Quantity:
5 g
25 g
100 g
71-27-2
C14H30Cl2N2O4
361.3
YOEWQQVKRJEPAE-UHFFFAOYSA-L
succinylcholine chloride, succinyldicholine chloride, suxamethonium chloride, midarine, scoline, succinylcholine dichloride, anectine, lysthenon, succinylforte, lysthenone
22475
CHEBI:61219
trimethyl-[2-[4-oxo-4-[2-(trimethylazaniumyl)ethoxy]butanoyl]oxyethyl]azanium;dichloride
C[N+](C)(C)CCOC(=O)CCC(=O)OCC[N+](C)(C)C.[Cl-].[Cl-]
71-27-2
C14H30Cl2N2O4
361.3
YOEWQQVKRJEPAE-UHFFFAOYSA-L
succinylcholine chloride, succinyldicholine chloride, suxamethonium chloride, midarine, scoline, succinylcholine dichloride, anectine, lysthenon, succinylforte, lysthenone
22475
CHEBI:61219
trimethyl-[2-[4-oxo-4-[2-(trimethylazaniumyl)ethoxy]butanoyl]oxyethyl]azanium;dichloride
C[N+](C)(C)CCOC(=O)CCC(=O)OCC[N+](C)(C)C.[Cl-].[Cl-]
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 160.0°C to 162.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (on dry substance) (Argentometry)
Quantity 5 g
Solubility Information Solubility in water: soluble.
Formula Weight 361.3
Percent Purity 96%
Chemical Name or Material Succinylcholine chloride
compliance-icons
Product Identifier
  • Succinylcholine chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /