Läs mer
Succinylcholine Chloride Dihydrate, TRC
Produktkod. 30416481
Change view
Kvantitet:
1 g
5 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30416481 5 g
30416491 50 g
30422080 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30416481 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6921035G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6101-15-1
Molekylformel C14 H30 N2 O4 . 2 Cl . 2 H2 O
Molekylvikt (g/mol) 397.34
Synonym Suxamethonium chloride Dihydrate, 2,2'-[butanedioylbis(oxy)]bis(N,N,N-trimethylethanaminium) dichloride, Succinylcholine Chloride Dihydrate, 2,2'-[(1,4-Dioxo-1,4-butanediyl)bis(oxy)]bis[N,N,N-trimethylethanaminium chloride hydrate (1:2:2), Choline chloride succinate (2:1) dihydrate, 2,2'-[(1,4-Dioxo-1,4-butanediyl)bis(oxy)]bis[N,N,N-trimethylethanaminium dichloride dihydrate, Bis[2-(dimethylamino)ethyl] succinate bismethochloride dihydrate, Choline succinate dichloride dihydrate, Succinic acid bis(2-dimethylaminoethyl) ester, dimethochloride, dihydrate, Succinylcholine chloride dihydrate, Succinylcholine chloride, 268U48
IUPAC-namn trimethyl-[2-[4-oxo-4-[2-(trimethylazaniumyl)ethoxy]butanoyl]oxyethyl]azanium;dichloride;dihydrate
LEDER O.O.[Cl-].[Cl-].C[N+](C)(C)CCOC(=O)CCC(=O)OCC[N+](C)(C)C
Specifikationer
Kemiskt namn eller material Suxamethonium chloride
Kvantitet 5 g
InChI formel InChI=1S/C14H30N2O4.2ClH.2H2O/c1-15(2,3)9-11-19-13(17)7-8-14(18)20-12-10-16(4,5)6;;;;/h7-12H2,1-6H3;2*1H;2*1H2/q+2;;;;/p-2
Formel vikt 396.1794