Läs mer
Sucrose-1-13Cfru, TRC
Produktkod. 30420489
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30420489 10 mg
1 options
Produktkod. 30420489 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS69700310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Sucrose-1-13Cfru
CAS 154368-11-3
Molekylformel 13CC11H22O11
Kvantitet 10 mg
Synonym β-D-Fructofuranosyl-13C α-D-Glucopyranoside
InChI formel InChI=1S/C12H22O11/c13-1-4-6(16)8(18)9(19)11(21-4)23-12(3-15)10(20)7(17)5(2-14)22-12/h4-11,13-20H,1-3H2/t4-,5-,6-,7-,8+,9-,10+,11-,12+/m1/s1/i3+1
LEDER O[C@H]1[C@H](O)[C@@H](O)[C@@H](O[C@@]2([13C]([H])([H])O)[C@@H](O)[C@H](O)[C@@H](CO)O2)O[C@@H]1CO
Molekylvikt (g/mol) 343.29
Formel vikt 343.1196