missing translation for 'onlineSavingsMsg'
Läs mer

Sucrose 6,1’,6’-Tricarboxylic Acid Trisodium Salt, TRC

Product Code. 30412561
Change view
Click to view available options
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30412561 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412561 Supplier Toronto Research Chemicals Supplier No. TRCS69707025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Sucrose 6,1',6'-Tricarboxylic Acid Trisodium Salt
CAS 226932-61-2
Molekylformel C12 H13 Na3 O14
Kvantitet 25 mg
Synonym β-D-Arabino-2-Hexulofuranosidaric acid','α-D-Glucopyranuronosyl Trisodium Salt
InChI formel InChI=1S/C12H16O14.3Na/c13-1-2(14)5(8(18)19)24-10(3(1)15)26-12(11(22)23)7(17)4(16)6(25-12)9(20)21;;;/h1-7,10,13-17H,(H,18,19)(H,20,21)(H,22,23);;;/q;3*+1/p-3/t1-,2-,3+,4+,5-,6-,7-,10+,12+;;;/m0.../s1
LEDER O[C@@H]1[C@@H](O)[C@@H](O[C@@]2(O[C@@H]([C@@H](O)[C@@H]2O)C(=O)O[Na])C(=O)O[Na])O[C@@H]([C@H]1O)C(=O)O[Na]
IUPAC-namn [(2S,3S,4S,5R,6R)-6-[(2R,3S,4S,5S)-3,4-dihydroxy-2,5-bis(sodiooxycarbonyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxane-2-carbonyl]oxysodium
Molekylvikt (g/mol) 450.192
Formel vikt 450
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.