Läs mer
Sucrose Brix/RI Standards, Paragon Scientific™
Produktkod. 16070410
Change view
Koncentration:
10,00 Brix
12,50 Brix
15,00 Brix
20,00 Brix
25,00 Brix
30,00 Brix
35 Brix
40,00 Brix
45,00 Brix
5,00 Brix
50,00 Brix
55 Brix
7,50 Brix
Förpackningsstorlek:
15ml
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Koncentration unitSize
16070410 20,00 Brix 15ml
16080400 5,00 Brix 15ml
16090400 7,50 Brix 15ml
16000410 10,00 Brix 15ml
16040410 12,50 Brix 15ml
16050410 15,00 Brix 15ml
16080410 25,00 Brix 15ml
16090410 30,00 Brix 15ml
16010420 35 Brix 15ml
16020420 40,00 Brix 15ml
16030420 45,00 Brix 15ml
16040420 50,00 Brix 15ml
16050420 55 Brix 15ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Produktkod. 16070410 Leverantör Paragon Scientific Leverantörsnummer SS20
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Paragon ISO 17025 and ISO 17034 certified brix standards are for the calibration and verification of all types of refractometer e.g handheld, Abbe and high accuracy digital instruments. Typically used within the food and beverage and pharmaceutical industries.
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Primary level refractive index standards traceable to National Standards
  • Manufactured in accordance with ICUMSA Methods
  • Supplied in tamper evident packs with 6 weeks shelf life
Specifikationer
Koncentration 20,00 Brix
Kvantitet 15 mL
Angivna nominella värden