missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sucrose, ( FCC), MP Biomedicals
Click to view available options
Quantity:
5 lb.
25 lb.
100 lb.
Specifications
Quantity | 5 lb. |
IUPAC Name | (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol |
Formula Weight | 342.3 |
Grade | FCC |
Chemical Name or Material | Sucrose |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'