Läs mer
Sucrose Octasulfate, Sodium Salt, TRC
Produktkod. 30410256
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30410256 500 mg
1 options
Produktkod. 30410256 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS699020500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Sucrose Octasulfate','Sodium Salt
CAS 74135-10-7
Molekylformel C12 H14 O35 S8 . 8 Na
Kvantitet 500 mg
Synonym α-D-Glucopyranoside','1,3,4,6-tetra-O-sulfo-β-D-fructofuranosyl','2,3,4,6-tetrakis(hydrogen sulfate)','sodium salt (1:8) (ACI), α-D-Glucopyranoside','1,3,4,6-tetra-O-sulfo-β-D-fructofuranosyl','tetrakis(hydrogen sulfate)','octasodium salt (9CI), Sodium sucrose octasulfate, Sucrose octasulfate sodium salt, Octasodium 2-(3,4-bis(sulfonatooxy)-2,5-bis(sulfonatooxymethyl)tetrahydrofuran-2-yloxy)-3,5-bis(sulfonatooxy)-6-(sulfonatooxymethyltetrahydropyran-4-oxysulfonate, 1,3,4,6-Tetra-O-sulfo-β-D-fructofuranosyl-α-D-glucopyranoside tetrakis(hydrogen sulfate) octasodium salt
InChI formel InChI=1S/C12H22O35S8.8Na/c13-48(14,15)37-1-4-6(43-51(22,23)24)8(45-53(28,29)30)9(46-54(31,32)33)11(40-4)42-12(3-39-50(19,20)21)10(47-55(34,35)36)7(44-52(25,26)27)5(41-12)2-38-49(16,17)18;;;;;;;;/h4-11H,1-3H2,(H,13,14,15)(H,16,17,18)(H,19,20,21)(H,22,23,24)(H,25,26,27)(H,28,29,30)(H,31,32,33)(H,34,35,36);;;;;;;;/q;8*+1/p-8/t4-,5-,6-,7-,8+,9-,10+,11-,12+;;;;;;;;/m1......../s1
LEDER [Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)OC[C@H]1O[C@H](O[C@]2(COS(=O)(=O)[O-])O[C@H](COS(=O)(=O)[O-])[C@@H](OS(=O)(=O)[O-])[C@@H]2OS(=O)(=O)[O-])[C@H](OS(=O)(=O)[O-])[C@@H](OS(=O)(=O)[O-])[C@@H]1OS(=O)(=O)[O-]
IUPAC-namn octasodium;[(2R,3R,4S,5R,6R)-2-[(2S,3S,4R,5R)-3,4-disulfonatooxy-2,5-bis(sulfonatooxymethyl)oxolan-2-yl]oxy-3,5-disulfonatooxy-6-(sulfonatooxymethyl)oxan-4-yl] sulfate
Molekylvikt (g/mol) 1158.66
Formel vikt 1157.6263