Läs mer
Sudoxicam, TRC
Produktkod. 30136006
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136006 1 g
30009952 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136006 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6889501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 34042-85-8
Molekylformel C13 H11 N3 O4 S2
Molekylvikt (g/mol) 337.37
Synonym 4-Hydroxy-2-methyl-N-2-thiazolyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide;
IUPAC-namn 4-hydroxy-2-methyl-1,1-dioxo-N-(1,3-thiazol-2-yl)-1λ^{6},2-benzothiazine-3-carboxamide
LEDER CN1C(=C(O)c2ccccc2S1(=O)=O)C(=O)Nc3nccs3
Specifikationer
Kemiskt namn eller material Sudoxicam
Kvantitet 1 g
InChI formel InChI=1S/C13H11N3O4S2/c1-16-10(12(18)15-13-14-6-7-21-13)11(17)8-4-2-3-5-9(8)22(16,19)20/h2-7,17H,1H3,(H,14,15,18)
Formel vikt 337.0191