Läs mer
Sulbentine, MedChemExpress
Produktkod. 16418636
Encompass_Preferred
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16418636 100 mg
16414557 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16418636 Leverantör Medchem Express Leverantörsnummer HYB1133
MedChemExpress Sulbentine (Dibenzthione) is an azole antifungal agent that has fungistatic and fungicidal activities. Sulbentine is used as a locally acting antimycotic in vivo.
Sulbentine (Dibenzthione) is an azole antifungal agent that has fungistatic and fungicidal activities. Sulbentine is used as a locally acting antimycotic in vivo.
Specifikationer
Kemiskt namn eller material Sulbentine
Fysisk form Solid
CAS 350-12-9
Färg White
Kvantitet 100 mg
Anteckningar om renhetsgrad Research
Molekylformel C17H18N2S2
För användning med (applikation) COVID-19-immunoregulation
Synonym Dibenzthione
Löslighetsinformation DMSO : 125 mg/mL (397.49 mM; Need ultrasonic)
LEDER S=C1SCN(CC2=CC=CC=C2)CN1CC3=CC=CC=C3
Molekylvikt (g/mol) 314.47
Formel vikt 314.47
Procent renhet 98.1%