Läs mer
Sulfabenzamide, MedChemExpress
Produktkod. 16472226
Encompass_Preferred
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
16472226 500 mg
1 options
Produktkod. 16472226 Leverantör Medchem Express Leverantörsnummer HYB0960
MedChemExpress Sulfabenzamide (N-Sulfanilylbenzamide) is an antimicrobial agent and usually consumed in combination with Sulfathiazole and Sulfacetamide. Sulfabenzamide is effective against Gram-positive and negative bacterial strains.
Sulfabenzamide (N-Sulfanilylbenzamide) is an antimicrobial agent and usually consumed in combination with Sulfathiazole and Sulfacetamide. Sulfabenzamide is effective against Gram-positive and negative bacterial strains.
Specifikationer
Kemiskt namn eller material Sulfabenzamide
Fysisk form Solid
CAS 127-71-9
Färg White
Kvantitet 500 mg
Anteckningar om renhetsgrad Research
Molekylformel C13H12N2O3S
För användning med (applikation) Cancer-programmed cell death
Synonym N-Sulfanilylbenzamide
Löslighetsinformation DMSO : 100 mg/mL (361.91 mM; Need ultrasonic)
LEDER O=C(NS(=O)(C1=CC=C(N)C=C1)=O)C2=CC=CC=C2
Molekylvikt (g/mol) 276.31
Formel vikt 276.31
Procent renhet 99.55%