missing translation for 'onlineSavingsMsg'
Läs mer

Sulfabenzamide, Mikromol™

Produktkod. 17954106
Change view
Click to view available options
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
17954106 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17954106 Supplier Mikromol Supplier No. MM3663.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Sulfabenzamide
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Sulfabenzamide
CAS 127-71-9
Analyt- eller komponentnamn Sulfabenzamide
Frakt skick Room Temperature
Molekylformel C13 H12 N2 O3 S
Certifikat och efterlevnad ISO 17025
Synonym Benzamide, N-[(4-aminophenyl)sulfonyl]-, Benzamide, N-sulfanilyl- (6CI,7CI,8CI), N-(Benzoyl)-4-aminobenzenesulfonamide, N-(p-Aminobenzenesulfonyl)benzamide, N-Sulfamylbenzamide, N-Sulfanilylbenzamide, N1-Benzoylsulfanilamide, NSC 74587, NSC 74587G, Sulfabenzamid, Sulfabenzamide, Sulfabenzid, Sulfabenzide, Sulfabenzoylamide
InChI formel InChI=1S/C13H12N2O3S/c14-11-6-8-12(9-7-11)19(17,18)15-13(16)10-4-2-1-3-5-10/h1-9H,14H2,(H,15,16)
LEDER Nc1ccc(cc1)S(=O)(=O)NC(=O)c2ccccc2
IUPAC-namn N-(4-aminophenyl)sulfonylbenzamide
Molekylvikt (g/mol) 276.31
Kvantitet 250 mg
Formel vikt 276.0569 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.