Läs mer
Sulfachloropyrazine Sodium Monohydrate, TRC
Produktkod. 30136086
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136086 1 g
17932855 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136086 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6889751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1392129-96-2
Molekylformel C10 H8 Cl N4 O2 S . Na . H2 O
Molekylvikt (g/mol) 324.72
Synonym Sulfaclozine Sodium Monohydrate, Benzenesulfonamide, 4-amino-N-(6-chloro-2-pyrazinyl)-, sodium salt, hydrate (1:1:1)
IUPAC-namn sodium;(4-aminophenyl)sulfonyl-(6-chloropyrazin-2-yl)azanide;hydrate
LEDER O.[Na+].Nc1ccc(cc1)S(=O)(=O)[N-]c2cncc(Cl)n2
Specifikationer
Kemiskt namn eller material Sulfaclozine sodium monohydrate
Kvantitet 1 g
InChI formel InChI=1S/C10H8ClN4O2S.Na.H2O/c11-9-5-13-6-10(14-9)15-18(16,17)8-3-1-7(12)2-4-8;;/h1-6H,12H2;;1H2/q-1;+1;
Formel vikt 324.006