Learn More
Sulfadiazine Silver Salt, TRC
Product Code. 30240756
Change view
Click to view available options
Quantity:
10 mg
10 g
25 g
Unit Size:
10g
10mg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240756 25 g 25g
30240867 10 mg 10mg
30240602 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30240756 Supplier Toronto Research Chemicals Supplier No. TRCS69905025G
 Restricted Item
Beräknad leverans: 03-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 22199-08-2
Molecular Formula C10 H9 N4 O2 S . Ag
Molecular Weight (g/mol) 357.14
Synonym Silver, [4-amino-N-(2-pyrimidinyl-κN1)benzenesulfonamidato-κO]-, Silver, (4-amino-N-2-pyrimidinylbenzenesulfonamidato-NN,O1)-, Sulfanilamide, N1-2-pyrimidinyl-, monosilver(1+) salt (8CI), Benzenesulfonamide, 4-amino-N-2-pyrimidinyl-, silver complex, Altreet-Ag, Dermazin, Dermazine, Flamazine, Flammazine, Geben, NSC 625324, Physiotulle-Ag, Silvadene, Silver 2-sulfanilamidopyrimidine, Silver sulfadiazine, Silver sulphadiazine, Silverol, Sulfadiazine silver, Sulfadiazine silver salt
IUPAC Name silver;(4-aminophenyl)sulfonyl-pyrimidin-2-ylazanide
SMILES [Ag+].Nc1ccc(cc1)S(=O)(=O)[N-]c2ncccn2
Specifications
Quantity 25 g
InChI Formula InChI=1S/C10H9N4O2S.Ag/c11-8-2-4-9(5-3-8)17(15,16)14-10-12-6-1-7-13-10;/h1-7H,11H2;/q-1;+1
Formula Weight 355.9497
Chemical Name or Material Silver Sulfadiazine