Läs mer
Sulfadimethoxine, TRC
Produktkod. 30138584
Change view
Kvantitet:
50 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138584 500 mg
30137822 1 g
17904385 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138584 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS699060500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 122-11-2
Molekylformel C12 H14 N4 O4 S
Molekylvikt (g/mol) 310.33
Synonym Benzenesulfonamide, 4-amino-N-(2,6-dimethoxy-4-pyrimidinyl)-, Sulfanilamide, N1-(2,6-dimethoxy-4-pyrimidinyl)- (6CI,8CI), 2,4-Dimethoxy-6-sulfanilamido-1,3-diazine, 2,4-Dimethoxy-6-sulfanilamido-1,3-pyrimidine, 2,4-Dimethoxy-6-sulfanilamidopyrimidine, 2,4-Dimethoxy-6-sulfonylamido-1,3-diazine, 2,6-Dimethoxy-4-sulfanilamidopyrimidine, 4-(p-Aminobenzenesulfonamido)-2,6-dimethoxypyrimidine, 4-Sulfa-2,6-dimethoxypyrimidine, 4-Sulfanilamido-2,6-dimethoxypyrimidine, 6-Sulfanilamido-2,4-dimethoxy-1,3-diazine, 6-Sulfanilamido-2,4-dimethoxypyrimidine, Abcid, Agribon, Albon, Arnosulfan, Bactrovet, Deposul, Diasulfa, Dimethoxysulfadiazine, Dinosol, Dorisul, Fuxal, Lasibon, Madribon, Madrigid, Madriqid, Madroxin, Madroxine, Maxulvet, Mecozine, Metoxidon, N-(2,6-Dimethoxypyrimidin-4-yl)-4-aminobenzenesulfonamide, N1-(2,6-Dimethoxy-4-pyrimidinyl)sulfanilamide, NSC 683544, Neostreptal, Omnibon, Persulfen, Redifal, SDMO, Scandisil, Sudine, Suldixine, Sulfabon, Sulfadimethoxin, Sulfadimethoxine, Sulfadimethoxydiazine, Sulfadimetoxin, Sulfadimetoxine, Sulfadimoxine, Sulfamed G, Sulfastop, Sulfoplan, Sulforal, Sulphadimethoxine, Sulxin, Sumbio, Symbio, Ultrasulfon
IUPAC-namn 4-amino-N-(2,6-dimethoxypyrimidin-4-yl)benzenesulfonamide
LEDER COc1cc(NS(=O)(=O)c2ccc(N)cc2)nc(OC)n1
Specifikationer
Kemiskt namn eller material Sulfadimethoxine
Kvantitet 500 mg
InChI formel InChI=1S/C12H14N4O4S/c1-19-11-7-10(14-12(15-11)20-2)16-21(17,18)9-5-3-8(13)4-6-9/h3-7H,13H2,1-2H3,(H,14,15,16)
Formel vikt 310.0736