Läs mer
Sulfadoxine, TRC
Produktkod. 30136126
Change view
Kvantitet:
100 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136126 1 g
30138565 10 g
17942525 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136126 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6990701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2447-57-6
Molekylformel C12 H14 N4 O4 S
Molekylvikt (g/mol) 310.33
Synonym Sulfadoxine, 4-amino-N-(5,6-dimethoxypyrimidin-4-yl)benzenesulfonamide, Benzenesulfonamide, 4-amino-N-(5,6-dimethoxy-4-pyrimidinyl)-, Sulfanilamide, N1-(5,6-dimethoxy-4-pyrimidinyl)-, 4,5-Dimethoxy-6-sulfanilamidopyrimidine, 4-Amino-N-(5,6-dimethoxy-4-pyrimidinyl)benzensulfonamide, 4-Sulfanilamido-5,6-dimethoxypyrimidine, Fanasil, Fanasulf, N1-5,6-Dimethoxy-4-pyrimidinylsulfanilamide, Orthosulfin, Ro 4-4393, Sanasil, Sulfadoxin, Sulfadoxine, Sulformethoxine, Sulformetoxin, Sulformetoxine, Sulforthodimethoxine, Sulforthomidine, WR 4073
IUPAC-namn 4-amino-N-(5,6-dimethoxypyrimidin-4-yl)benzenesulfonamide
LEDER COc1ncnc(NS(=O)(=O)c2ccc(N)cc2)c1OC
Specifikationer
Kemiskt namn eller material Sulfadoxine
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C12H14N4O4S/c1-19-10-11(14-7-15-12(10)20-2)16-21(17,18)9-5-3-8(13)4-6-9/h3-7H,13H2,1-2H3,(H,14,15,16)
Formel vikt 310.0736
Procent renhet >95%