Läs mer
Sulfaethoxypyridazine, TRC
Produktkod. 30008909
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008909 100 mg
30007030 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008909 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS699255100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 963-14-4
Molekylformel C12 H14 N4 O3 S
Molekylvikt (g/mol) 294.33
Synonym Sulfanilamide, N1-(6-ethoxy-3-pyridazinyl)- (6CI,8CI), Sulfanilamide, N1-(6-ethoxy-3-pyridazinyl)- (7CI), 4-Amino-N-(6-ethoxy-3-pyridazinyl)benzenesulfonamide, Cysul, Sulfaethoxypyridazine, Sulfanilamido-6-ethoxy-3-pyridazine
IUPAC-namn 4-amino-N-(6-ethoxypyridazin-3-yl)benzenesulfonamide
LEDER CCOc1ccc(NS(=O)(=O)c2ccc(N)cc2)nn1
Specifikationer
Kemiskt namn eller material Sulfaethoxypyridazine
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C12H14N4O3S/c1-2-19-12-8-7-11(14-15-12)16-20(17,18)10-5-3-9(13)4-6-10/h3-8H,2,13H2,1H3,(H,14,16)
Formel vikt 294.0787
Procent renhet >95%