Läs mer
Sulfamethazine-13C6, TRC
Produktkod. 17964065
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17964065 1 mg
17924955 5 mg
17943635 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17964065 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6990731MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 77643-91-5
Molekylformel 13C6 C6 H14 N4 O2 S
Molekylvikt (g/mol) 284.29
Synonym Benzene-13C6-sulfonamide, 4-amino-N-(4,6-dimethyl-2-pyrimidinyl)- (9CI), Sulfamethazine (phenyl-13C6), Sulfadimidine (phenyl-13C6)
IUPAC-namn 4-amino-N-(4,6-dimethylpyrimidin-2-yl)(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-triene-1-sulfonamide
LEDER Cc1cc(C)nc(NS(=O)(=O)[13c]2[13cH][13cH][13c](N)[13cH][13cH]2)n1
Specifikationer
Kemiskt namn eller material Sulfamethazine phenyl-13C6
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C12H14N4O2S/c1-8-7-9(2)15-12(14-8)16-19(17,18)11-5-3-10(13)4-6-11/h3-7H,13H2,1-2H3,(H,14,15,16)/i3+1,4+1,5+1,6+1,10+1,11+1
Formel vikt 284.1039
Procent renhet >95%