Läs mer
Sulfamethizole, TRC
Produktkod. 30418457
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30418457 10 g
30415837 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30418457 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS68900510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 144-82-1
Molekylformel C9 H10 N4 O2 S2
Molekylvikt (g/mol) 270.3313
Synonym Sulfamethizole, 4-amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)benzenesulfonamide, Benzenesulfonamide, 4-amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)-, Sulfanilamide, N1-(5-methyl-1,3,4-thiadiazol-2-yl)- (6CI,7CI,8CI), 2-Methyl-5-sulfanilamido-1,3,4-thiadiazole, 2-Sulfanilamido-5-methyl-1,3,4-thiadiazole, 4-Amino-N-(5-methyl-[1,3,4]thiadiazol-2-yl)benzenesulfonamide, 5-Methyl-2-sulfanilamido-1,3,4-thiadiazole, Ayerlucil, Famet, Lucosil, Methazol, Microsul, N-(5-Methyl-1,3,4-thiadiazol-2-yl)-4-aminobenzenesulfonamide, N1-(5-Methyl-1,3,4-thiadiazol-2-yl)sulfanilamide, Renasul, Rufol, Salimol, Sulfamethiazole, Sulfamethizol, Sulfamethizole, Sulfamethylthiadiazole, Sulfapyelon, Sulfstat, Sulfurine, Sulphamethizole, Tetracid, Thidicur, Thiosulfil, Thiosulfil Forte, Ultrasul, Urocydal, Urodiaton, Urolucosil, Urosulfin, VK 53
IUPAC-namn 4-amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)benzenesulfonamide
LEDER Cc1nnc(NS(=O)(=O)c2ccc(N)cc2)s1
Specifikationer
Kemiskt namn eller material Sulfamethizole
Anteckningar om renhetsgrad HPLC
Kvantitet 10 g
InChI formel InChI=1S/C9H10N4O2S2/c1-6-11-12-9(16-6)13-17(14,15)8-4-2-7(10)3-5-8/h2-5H,10H2,1H3,(H,12,13)
Formel vikt 270.0245
Procent renhet >95%