Läs mer
Sulfamethoxazole Beta-D-Glucuronide, TRC
Produktkod. 30003802
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30003802 10 mg
30005685 1 mg
30009186 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30003802 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS69913510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14365-52-7
Molekylformel C16 H19 N3 O9 S
Molekylvikt (g/mol) 429.4
Synonym Sulfamethoxazole B-D-Glucuronide
IUPAC-namn (2S,3S,4S,5R,6R)-6-[(4-aminophenyl)sulfonyl-(5-methyl-1,2-oxazol-3-yl)amino]-3,4,5-trihydroxyoxane-2-carboxylic acid
LEDER Cc1onc(c1)N([C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O)S(=O)(=O)c3ccc(N)cc3
Specifikationer
Kemiskt namn eller material Sulfamethoxazole beta-D-Glucuronide
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C16H19N3O9S/c1-7-6-10(18-28-7)19(29(25,26)9-4-2-8(17)3-5-9)15-13(22)11(20)12(21)14(27-15)16(23)24/h2-6,11-15,20-22H,17H2,1H3,(H,23,24)/t11-,12-,13+,14-,15+/m0/s1
Formel vikt 429.0842
Procent renhet >95%