missing translation for 'onlineSavingsMsg'
Learn More

Sulfamethoxypyridazine, Thermo Scientific Chemicals

Catalog Number p-4229330
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
80-35-3
C11H12N4O3S
280.3
VLYWMPOKSSWJAL-UHFFFAOYSA-N
sulfamethoxypyridazine, sulphamethoxypyridazine, sulfapyridazine, sulfalex, longin, midicel, sulfametoxipiridazine, spofadazine, sulfdurazin, 3-sulfa-6-methoxypyridazine
5330
CHEBI:102516
4-amino-N-(6-methoxypyridazin-3-yl)benzenesulfonamide
COC1=NN=C(C=C1)NS(=O)(=O)C2=CC=C(C=C2)N
This item is not returnable. View return policy
80-35-3
C11H12N4O3S
280.3
VLYWMPOKSSWJAL-UHFFFAOYSA-N
sulfamethoxypyridazine, sulphamethoxypyridazine, sulfapyridazine, sulfalex, longin, midicel, sulfametoxipiridazine, spofadazine, sulfdurazin, 3-sulfa-6-methoxypyridazine
5330
CHEBI:102516
4-amino-N-(6-methoxypyridazin-3-yl)benzenesulfonamide
COC1=NN=C(C=C1)NS(=O)(=O)C2=CC=C(C=C2)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 182°C to 183°C
Color White to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 g
Merck Index 15,9051
Solubility Information Solubility in water: insoluble
Formula Weight 280.3
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material Sulfamethoxypyridazine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sulfamethoxypyridazine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'