Läs mer
Sulfamonomethoxine Sodium, TRC
Produktkod. 30415151
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30415151 50 mg
30410215 100 mg
30421342 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30415151 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS69927850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 38006-08-5
Molekylformel C11 H11 N4 O3 S . Na
Molekylvikt (g/mol) 302.28
Synonym Benzenesulfonamide, 4-amino-N-(6-methoxy-4-pyrimidinyl)-, monosodium salt (9CI), Sodium sulfamonomethoxine, Sulfamonomethoxine sodium, Sulfamonomethoxine sodium salt
IUPAC-namn sodium;(4-aminophenyl)sulfonyl-(6-methoxypyrimidin-4-yl)azanide
LEDER [Na+].COc1cc([N-]S(=O)(=O)c2ccc(N)cc2)ncn1
Specifikationer
Kemiskt namn eller material Sulfamonomethoxine sodium
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C11H11N4O3S.Na/c1-18-11-6-10(13-7-14-11)15-19(16,17)9-4-2-8(12)3-5-9;/h2-7H,12H2,1H3;/q-1;+1
Formel vikt 302.045
Procent renhet >95%