Learn More
Sulfanilic acid, Sigma-Aldrich™
Specifications
Specifications
| Chemical Name or Material | Sulfanilic acid |
| CAS | 121-57-3 |
| Melting Point | 288°C (decomposes) |
| Density | 1.485 g/cm³ at 25°C |
| Quantity | 50 kg |
| Synonym | Sulfanilic acid, 4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid, Sulfanilic acid |
| Molecular Weight (g/mol) | 173.19 |
| Grade | Not Available |