Läs mer
Sulfanilsyra natriumsalt, MP Biomedicals™
Produktkod. 11495131 Handla allt MP Biomedicals Produkter
Change view
Kvantitet:
50 g
250 g
Förpackningsstorlek:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11495131 50 g 50g
11405141 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11495131 Leverantör MP Biomedicals Leverantörsnummer 0215671650
Kemiska identifierare
CAS 6106-22-5
Molekylformel C6H10NNaO5S
Molekylvikt (g/mol) 231.198
InChI-nyckel MJFZIDUBUNIFLZ-UHFFFAOYSA-M
Synonym sodium sulfanilate dihydrate, unii-y31oen1192, 4-aminobenzenesulfonic acid, sodium salt dihydrate, sodium 4-aminobenzenesulfonate dihydrate, benzenesulfonic acid,4-amino-, sodium salt, hydrate 1:1:?, sulfanilic acid sodium salt dihydrate mi, acmc-1b9i8, c6h6nso3.na.2h2o, benzenesulfonic acid, 4-amino-, sodium salt, hydrate 1:1:2, 4h-1,4-benzoxazin-4-amine,2,3-dihydro hcl
PubChem CID 23687713
IUPAC-namn natrium;4-aminobensensulfonat;dihydrat
LEDER C1=CC(=CC=C1N)S(=O)(=O)[O-].O.O.[Na+]
Specifikationer
Kemiskt namn eller material Sulfanilic acid sodium salt
Färg Vitt
Kvantitet 50 g
Formel vikt 195.2
Fysisk form Pulver