Läs mer
Sulfaphenazole, TRC
Produktkod. 30422816
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30422816 250 mg
1 options
Produktkod. 30422816 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS689020250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Sulfaphenazole
CAS 526-08-9
Anteckningar om renhetsgrad HPLC
Molekylformel C15 H14 N4 O2 S
Kvantitet 250 mg
Synonym Benzenesulfonamide','4-amino-N-(1-phenyl-1H-pyrazol-5-yl)-, Sulfanilamide','N1-(1-phenylpyrazol-5-yl)- (8CI), 1-Phenyl-5-sulfanilamidopyrazole, 2-Phenyl-3-sulfanilamidopyrazole, 3-(p-Aminobenzenesulfonamido)-2-phenylpyrazole, 5-Sulfanilamido-1-phenylpyrazole, Depocid, Depotsulfonamide, Eftolon, Firmazolo, Inamil, Isarol, Isarol V, Merian, Microtan pirazolo, N1-(1-Phenylpyrazol-5-yl)sulfanilamide, Orisul, Orisulf, Paidazolo, Plisulfan, Raziosulfa, Sulfabid, Sulfaphenazol, Sulfaphenazole, Sulfaphenazon, Sulfaphenylpyrazole, Sulphaphenazole
InChI formel InChI=1S/C15H14N4O2S/c16-12-6-8-14(9-7-12)22(20,21)18-15-10-11-17-19(15)13-4-2-1-3-5-13/h1-11,18H,16H2
LEDER Nc1ccc(cc1)S(=O)(=O)Nc2ccnn2c3ccccc3
IUPAC-namn 4-amino-N-(2-phenylpyrazol-3-yl)benzenesulfonamide
Molekylvikt (g/mol) 314.36
Formel vikt 314.0837