Läs mer
Sulfaquinoxaline Sodium Salt, TRC
Produktkod. 17963565
Change view
Kvantitet:
100 mg
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17963565 5 g
17941875 100 mg
17930855 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17963565 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6890365G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 967-80-6
Molekylformel C14 H11 N4 O2 S . Na
Molekylvikt (g/mol) 322.32
Synonym Sulfaquinoxaline Sodium
IUPAC-namn sodium;(4-aminophenyl)sulfonyl-quinoxalin-2-ylazanide
LEDER [Na+].Nc1ccc(cc1)S(=O)(=O)[N-]c2cnc3ccccc3n2
Specifikationer
Kemiskt namn eller material Sulfaquinoxaline sodium salt
Anteckningar om renhetsgrad HPLC
Kvantitet 5 g
InChI formel InChI=1S/C14H11N4O2S.Na/c15-10-5-7-11(8-6-10)21(19,20)18-14-9-16-12-3-1-2-4-13(12)17-14;/h1-9H,15H2;/q-1;+1
Formel vikt 322.05
Procent renhet >95%