Läs mer
Sulfate Standard: SO4(-2) @ 10000 μg/mL in H2O, VHG™
Produktkod. 17959154
Change view
Kvantitet:
100 ml
500 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17959154 500 ml
17999714 100 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17959154 Leverantör VHG Labs Leverantörsnummer VHGI1PSO4500
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Sulfate Standard: SO4(-2) @ 10000 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Sulfate
CAS 14808-79-8
Metoder Ion Chromatography/Wet Chemistry
Analyt- eller komponentnamn Sulfate
Molekylformel (S O4)2-
Certifikat och efterlevnad ISO 17034
Synonym SO42-
LEDER [O-]S(=O)(=O)[O-]
IUPAC-namn Sulfate
Lösningsmedel eller matris Water
Molekylvikt (g/mol) 96.06
Kvantitet 500 ml
Formel vikt 96.06 g/mol