Läs mer
Sulfatroxazole, TRC
Produktkod. 30138582
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138582 500 mg
30010433 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138582 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS699265500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23256-23-7
Molekylformel C11 H13 N3 O3 S
Molekylvikt (g/mol) 267.3
Synonym Sulfatroxazolum, 4-Amino-N-(3,4-dimethyl-5-isoxazolyl)benzenesulfonamide
IUPAC-namn 4-amino-N-(4,5-dimethyl-1,2-oxazol-3-yl)benzenesulfonamide
LEDER Cc1onc(NS(=O)(=O)c2ccc(N)cc2)c1C
Specifikationer
Kemiskt namn eller material Sulfatroxazole
Kvantitet 500 mg
InChI formel InChI=1S/C11H13N3O3S/c1-7-8(2)17-13-11(7)14-18(15,16)10-5-3-9(12)4-6-10/h3-6H,12H2,1-2H3,(H,13,14)
Formel vikt 267.0678