Läs mer
Sulfluramid (>90%), TRC
Produktkod. 30415220
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30415220 50 mg
1 options
Produktkod. 30415220 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS69909050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Sulfluramid
CAS 4151-50-2
Molekylformel C10 H6 F17 N O2 S
Kvantitet 50 mg
Synonym Volcano, N-Ethyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-1-octanesulfonamide (ACI), AI 3-29757, Alstar, Alstar (pesticide), FiniTron, FX 12, GX 071, Mirex S, N-Ethyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane-1-sulfonamide, N-Ethylperfluoro-1-octanesulfonamide, N-Ethylperfluorooctanesulfonamide, N-Ethylperfluorooctylsulfonamide, Sulfluramid, Sulfluramide, Volcano, Volcano (insecticide), 1-Octanesulfonamide','N-ethyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro- (7CI','8CI','9CI','ACI), EtFOSA
InChI formel InChI=1S/C10H6F17NO2S/c1-2-28-31(29,30)10(26,27)8(21,22)6(17,18)4(13,14)3(11,12)5(15,16)7(19,20)9(23,24)25/h28H,2H2,1H3
LEDER CCNS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC-namn N-ethyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane-1-sulfonamide
Molekylvikt (g/mol) 527.2
Formel vikt 526.9848