Läs mer
Sulfosuccinimidyl Oleate Sodium, TRC
Produktkod. 30008843
Change view
Kvantitet:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008843 25 mg
30006166 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008843 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS68920025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 135661-44-8
Molekylformel C22 H36 N O7 S . Na
Molekylvikt (g/mol) 481.58
IUPAC-namn sodium;1-[(Z)-octadec-9-enoyl]oxy-2,5-dioxopyrrolidine-3-sulfonate
LEDER [Na+].CCCCCCCC\C=C/CCCCCCCC(=O)ON1C(=O)CC(C1=O)S(=O)(=O)[O-]
Specifikationer
Kemiskt namn eller material Sulfosuccinimidyl Oleate Sodium
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C22H37NO7S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(25)30-23-20(24)18-19(22(23)26)31(27,28)29;/h9-10,19H,2-8,11-18H2,1H3,(H,27,28,29);/q;+1/p-1/b10-9-;
Formel vikt 481.211
Procent renhet >95%