Läs mer
Sulfoxaflor, TRC
Produktkod. 17942745
Change view
Kvantitet:
2.5 mg
10 mg
100 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17942745 2.5 mg 2.5mg
17904575 100 mg -
17911655 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17942745 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS7550052.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 946578-00-3
Molekylformel C10 H10 F3 N3 O S
Molekylvikt (g/mol) 277.27
Synonym N-[Methyloxido[1-[6-(trifluoromethyl)-3-pyridinyl]ethyl]-λ4-sulfanylidene]cyanamide, Closer, Closer SC, Fengchongding, GF 2032, GF 2372, Isoclast active, Methyl[1-(2-trifluoromethylpyridin-5-yl)ethyl]-N-cyanosulfoximine, Sulfoxaflor, Transform, XDE 208
IUPAC-namn [methyl-oxo-[1-[6-(trifluoromethyl)pyridin-3-yl]ethyl]-λ^{6}-sulfanylidene]cyanamide
LEDER CC(c1ccc(nc1)C(F)(F)F)S(=O)(=NC#N)C
Specifikationer
Kemiskt namn eller material Sulfoxaflor
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C10H10F3N3OS/c1-7(18(2,17)16-6-14)8-3-4-9(15-5-8)10(11,12)13/h3-5,7H,1-2H3
Formel vikt 277.0497
Procent renhet >95%