Learn More
Sulfur Standard: S @ 50 μg/g (0.0050 wt%) in 75 cSt Mineral Oil, VHG™
Product Code. 17960686
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17960686 500 mL 500mL
17951986 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17960686 Supplier VHG Labs Supplier No. VHGSMIN50500
 Restricted Item
Beräknad leverans: 10-12-2026
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Sulfur Standard: S @ 50 μg/g (0.0050 wt%) in 75 cSt Mineral Oil
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sulfur
CAS 7704-34-9
Methods XRF
Analyte or Component Names Sulfur
Shipping Condition Room Temperature
Molecular Formula S
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/S
SMILES [S]
IUPAC Name sulfur
Solvent or Matrix 75 cSt Mineral Oil
Molecular Weight (g/mol) 32.065
Quantity 500 mL
Formula Weight 31.972 g/mol