missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, 1.0N Standardized Solution, Thermo Scientific Chemicals

Catalog Number p-4408601
Click to view available options
Quantity:
1 L
4 L
20 L
7664-93-9, 7732-18-5
H2O4S
98.07
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
CHEBI:26836
sulfuric acid
OS(O)(=O)=O
7664-93-9, 7732-18-5
H2O4S
98.07
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
CHEBI:26836
sulfuric acid
OS(O)(=O)=O

Sulfuric acid, 1.0N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 L
Formula Weight 98.08
Concentration or Composition (by Analyte or Components) Sulfuric acid: 5%; Water: 95%
Solubility Miscible with water
Physical Form Liquid
Chemical Name or Material Sulfuric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'