Läs mer
Sulindac Acyl-Beta-D-Glucuronide, TRC
Produktkod. 30425257
Change view
Kvantitet:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425257 2.5 mg
30419370 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30425257 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6992452.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 60018-36-2
Molekylformel C26H25FO9S
Molekylvikt (g/mol) 532.53
Synonym (Z)-1-[5-Fluoro-2-methyl-1-[[4-(methylsulfinyl)phenyl]methylene]-1H-indene-3-acetate]β-D-Glucopyranuronic Acid
LEDER CC\1=C(C2=C(/C1=C\C3=CC=C(C=C3)S(=O)C)C=CC(=C2)F)CC(=O)O[C@H]4[C@@H]([C@H]([C@@H]([C@H](O4)C(=O)O)O)O)O
Specifikationer
Kemiskt namn eller material Sulindac Acyl-beta-D-Glucuronide
Kvantitet 2.5 mg
Formel vikt 532.1203