Läs mer
Sulindac Sulfide Methyl Ester, TRC
Produktkod. 30414903
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30414903 100 mg
1 options
Produktkod. 30414903 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS699315100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Sulindac Sulfide Methyl Ester
CAS 106647-49-8
Molekylformel C21H19FO2S
Kvantitet 100 mg
Synonym (1Z)-5-Fluoro-2-methyl-1-[[4-(methylthio)phenyl]methylene]-1H-indene-3-acetic Acid Methyl Ester, (Z)-5-Fluoro-2-methyl-1-[[4-(methylthio)phenyl]methylene]-1H-indene-3-acetic Acid Methyl Ester
InChI formel InChI=1S/C21H19FO2S/c1-13-18(10-14-4-7-16(25-3)8-5-14)17-9-6-15(22)11-20(17)19(13)12-21(23)24-2/h4-11H,12H2,1-3H3/b18-10-
LEDER COC(=O)CC1=C(C)\C(=C\c2ccc(SC)cc2)\c3ccc(F)cc13
IUPAC-namn methyl 2-[(3Z)-6-fluoro-2-methyl-3-[(4-methylsulfanylphenyl)methylidene]inden-1-yl]acetate
Molekylvikt (g/mol) 354.44
Formel vikt 354.109