Läs mer
Sulindac Sulfone Methyl Ester, TRC
Produktkod. 30422445
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30422445 250 mg
1 options
Produktkod. 30422445 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS699320250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Sulindac Sulfone Methyl Ester
CAS 141741-24-4
Molekylformel C21H19FO4S
Kvantitet 250 mg
Synonym (Z)-5-Fluoro-2-methyl-1-[[4-(methylsulfonyl)phenyl]methylene]-1H-indene-3-acetic Acid Methyl Ester, (1Z)-5-Fluoro-2-methyl-1[[4-(methylsulfonyl)-1H-indene-3-acetic Acid Methyl Ester
InChI formel InChI=1S/C21H19FO4S/c1-13-18(10-14-4-7-16(8-5-14)27(3,24)25)17-9-6-15(22)11-20(17)19(13)12-21(23)26-2/h4-11H,12H2,1-3H3/b18-10-
LEDER COC(=O)CC1=C(C)\C(=C\c2ccc(cc2)S(=O)(=O)C)\c3ccc(F)cc13
IUPAC-namn methyl 2-[(3Z)-6-fluoro-2-methyl-3-[(4-methylsulfonylphenyl)methylidene]inden-1-yl]acetate
Molekylvikt (g/mol) 386.44
Formel vikt 386.0988