Läs mer
Sulpiride EP Impurity E, Mikromol™
Produktkod. 30049376
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049376 25 mg
30051420 100 mg
30459196 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30049376 Leverantör Mikromol Leverantörsnummer MM0051.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Sulpiride EP Impurity E
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Methoxy-5-sulfamoylbenzamide
CAS 52395-25-2
Analyt- eller komponentnamn 2-Methoxy-5-sulfamoylbenzamide
Frakt skick Room Temperature
Molekylformel C8 H10 N2 O4 S
Certifikat och efterlevnad ISO 17025
Synonym 2-Methoxy-5-sulphamoylbenzamide, Sulpiride Imp. E (EP), Sulpiride Impurity E
InChI formel InChI=1S/C8H10N2O4S/c1-14-7-3-2-5(15(10,12)13)4-6(7)8(9)11/h2-4H,1H3,(H2,9,11)(H2,10,12,13)
LEDER COc1ccc(cc1C(=O)N)S(=O)(=O)N
IUPAC-namn 2-methoxy-5-sulfamoylbenzamide
Molekylvikt (g/mol) 230.24
Kvantitet 25 mg
Formel vikt 230.0361 g/mol