Learn More
SULT1B1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA551683
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KIEQFHSRPD DIVIATYPKS GTTWVSEIID MILNDGDIEK CKRGFITEKV PMLEMTLPGL RTSGIEQLEK NPSPRIVKTH LPTDLLPKSF WENNCKMIYL ARN Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 76%.
SULT1B1 Polyclonal antibody specifically detects SULT1B1 in Human samples. It is validated for Immunohistochemistry.Specifications
SULT1B1 | |
Polyclonal | |
Unconjugated | |
SULT1B1 | |
DOPA/tyrosine sulfotransferase; ST1B1; ST1B2; St2b2; sulfotransferase 1B1; sulfotransferase 1B2; sulfotransferase family 1B member 1; sulfotransferase family 1B, member 1; sulfotransferase family cytosolic 1B member 1; sulfotransferase family, cytosolic, 1B, member 1; SULT1B1; SULT1B2; Thyroid hormone sulfotransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27284 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43704 | |
SULT1B1 | |
Recombinant protein corresponding to Human SULT1B1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only