Läs mer
Sultamicillin Tosylate, TRC
Produktkod. 30423876
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30423876 250 mg
1 options
Produktkod. 30423876 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS789490250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Sultamicillin tosilate
CAS 83105-70-8
Molekylformel C25 H30 N4 O9 S2 . C7 H8 O3 S
Kvantitet 250 mg
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid','6-[[(2R)-aminophenylacetyl]amino]-3,3-dimethyl-7-oxo-','[[[(2S,6R)-3,3-dimethyl-4,4-dioxido-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-2-yl]carbonyl]oxy]methyl ester','(2S,5R,6R)-','(4-methylbenzenesulfonate) (1:1), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid','6-[(aminophenylacetyl)amino]-3,3-dimethyl-7-oxo-','[[(3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-2-yl)carbonyl]oxy]methyl ester','S,S-dioxide','[2S-[2α(2R*,5S*),5α,6β(S*)]]-','mono(4-methylbenzenesulfonate), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid','6-[[(2R)-aminophenylacetyl]amino]-3,3-dimethyl-7-oxo-','[[[(2S,6R)-3,3-dimethyl-4,4-dioxido-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-2-yl]carbonyl]oxy]methyl ester','(2S,5R,6R)-','mono(4-methylbenzenesulfonate) (9CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid','6-[(aminophenylacetyl)amino]-3,3-dimethyl-7-oxo-','[[(3,3-dimethyl-4,4-dioxido-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-2-yl)carbonyl]oxy]methyl ester','[2S-[2α(2R*,5S*),5α,6β(S*)]]-','mono(4-methylbenzenesulfonate), Bacimex, Bethacil orale, Sultamicillin p-toluenesulfonate, Sultamicillin tosylate, Unacid PD oral, Unacim orale
InChI formel InChI=1S/C25H30N4O9S2.C7H8O3S/c1-24(2)17(29-20(32)16(21(29)39-24)27-19(31)15(26)12-8-6-5-7-9-12)22(33)37-11-38-23(34)18-25(3,4)40(35,36)14-10-13(30)28(14)18;1-6-2-4-7(5-3-6)11(8,9)10/h5-9,14-18,21H,10-11,26H2,1-4H3,(H,27,31);2-5H,1H3,(H,8,9,10)/t14-,15-,16-,17+,18+,21-;/m1./s1
LEDER Cc1ccc(cc1)S(=O)(=O)O.CC2(C)S[C@@H]3[C@H](NC(=O)[C@H](N)c4ccccc4)C(=O)N3[C@H]2C(=O)OCOC(=O)[C@@H]5N6[C@@H](CC6=O)S(=O)(=O)C5(C)C
IUPAC-namn [(2S,5R)-3,3-dimethyl-4,4,7-trioxo-4λ^{6}-thia-1-azabicyclo[3.2.0]heptane-2-carbonyl]oxymethyl (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;4-methylbenzenesulfonic acid
Molekylvikt (g/mol) 766.86
Formel vikt 766.1648