Läs mer
Sultopride hydrochloride, MedChemExpress
Produktkod. 16493967
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16493967 50 mg
16450518 5 mg
16408058 10 mg
16403756 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16493967 Leverantör Medchem Express Leverantörsnummer HY42849A
MedChemExpress Sultopride hydrochloride (LIN-1418 hydrochloride) is a selective antagonist of dopamine D2 receptor.
Sultopride hydrochloride (LIN-1418 hydrochloride) is a selective antagonist of dopamine D2 receptor.
Specifikationer
Kemiskt namn eller material Sultopride hydrochloride
CAS 23694-17-9
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C17H27ClN2O4S
Kvantitet 50 mg
Synonym LIN-1418 hydrochloride
Löslighetsinformation H2O : 50 mg/mL (127.90 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (127.90 mM; Need ultrasonic)
LEDER O=C(NCC1N(CC)CCC1)C2=CC(S(=O)(CC)=O)=CC=C2OC.Cl
Molekylvikt (g/mol) 390.93
Formel vikt 390.93
Procent renhet 99.27%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation