Läs mer
Sumatriptan Hydroxy-Oxindole Impurity (Sumatriptan Impurity 1), TRC
Produktkod. 30426300
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30426300 5 mg
1 options
Produktkod. 30426300 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS8100055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Sumatriptan Hydroxy-Oxindole Impurity (Sumatriptan Impurity 1)
CAS 2250254-19-2
Molekylformel C14H21N3O4S
Kvantitet 5 mg
Synonym 1-(3-(2-(Dimethylamino)ethyl)-3-hydroxy-2-oxoindolin-5-yl)-N-methylmethane sulfonamide
InChI formel InChI=1S/C14H21N3O4S/c1-15-22(20,21)9-10-4-5-12-11(8-10)14(19,13(18)16-12)6-7-17(2)3/h4-5,8,15,19H,6-7,9H2,1-3H3,(H,16,18)
LEDER CNS(CC1=CC(C(CCN(C)C)(O)C(N2)=O)=C2C=C1)(=O)=O
Molekylvikt (g/mol) 327.4