Läs mer
Sumatriptan N-Oxide, TRC
Produktkod. 30135509
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135509 25 mg
30138685 50 mg
17983755 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135509 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS81001025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 212069-94-8
Molekylformel C14 H21 N3 O3 S
Molekylvikt (g/mol) 311.4
Synonym 3-[2-(Dimethyloxidoamino)ethyl]-N-methyl-1H-indole-5-methanesulfonamide, GR 112504, Sumatriptan N-Oxide, GR 112504X, Ph Eur Sumatriptan Impurity D, BP Sumatriptan Impurity D, Sumatriptan N-oxide (GR 112504X), Ph Eur Sumatriptan Impurity Mixture, Sumatriptan Test Mix for Impurity Identification Solution 2
IUPAC-namn N,N-dimethyl-2-[5-(methylsulfamoylmethyl)-1H-indol-3-yl]ethanamine oxide
LEDER CNS(=O)(=O)Cc1ccc2[nH]cc(CC[N+](C)(C)[O-])c2c1
Specifikationer
Kemiskt namn eller material Sumatriptan N-Oxide
Kvantitet 25 mg
InChI formel InChI=1S/C14H21N3O3S/c1-15-21(19,20)10-11-4-5-14-13(8-11)12(9-16-14)6-7-17(2,3)18/h4-5,8-9,15-16H,6-7,10H2,1-3H3
Formel vikt 311.1304