Learn More
SUMO2/3 Mouse anti-Human, Unconjugated, Clone: 1A1B3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67154-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The SUMO2/3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SUMO2/3. This antibody recognizes human, mouse, rat antigen. The SUMO2/3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
SUMO2/3 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
1A1B3 | |
Unconjugated | |
150 μL | |
6613 | |
IgG2b |