Läs mer
Tocris Bioscience™ SUN-B 8155
GREENER_CHOICE
Produktkod. 18044302
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18044302 10 mg
18095992 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18044302 Leverantör Tocris Bioscience™ Leverantörsnummer 2550/10
Non-peptide calcitonin (CT) receptor agonist
Non-peptide calcitonin (CT) receptor agonist that fully stimulates cAMP production in CHO/hCTR cells (EC50 = 21 μM). Causes a significant reduction in serum calcium concentrations in vivo following i.p. administration.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 345893-91-6
Molekylformel C14H15N3O3
Specifikationer
Kemiskt namn eller material SUN-B 8155
Kvantitet 10 mg
Formel vikt 273.29
Renhet >98%
Molekylvikt (g/mol) 273.292