Learn More
Tocris Bioscience™ SUN-B 8155
GREENER_CHOICE
Product Code. 18095992
Change view
Click to view available options
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
18095992 50 mg
18044302 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18095992 Supplier Tocris Bioscience™ Supplier No. 2550/50
Non-peptide calcitonin (CT) receptor agonist
Non-peptide calcitonin (CT) receptor agonist that fully stimulates cAMP production in CHO/hCTR cells (EC50 = 21 μM). Causes a significant reduction in serum calcium concentrations in vivo following i.p. administration.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 345893-91-6
Molekylformel C14H15N3O3
Specifications
Kemiskt namn eller material SUN-B 8155
Kvantitet 50 mg
Formel vikt 273.29
Renhet >98%
Molekylvikt (g/mol) 273.292