missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUN1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24568-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene is a member of the unc-84 homolog family and encodes a nuclear envelope protein with an Unc84 (SUN) domain. The protein is involved in nuclear anchorage and migration. Several alternatively spliced transcript variants of this gene have been described; however, the full-length nature of some of these variants has not been determined.Specifications
SUN1 | |
Polyclonal | |
Unconjugated | |
SUN1 | |
KIAA0810, Protein unc 84 homolog A, Sad1/unc 84 protein like 1, SUN1, unc 84 homolog A (C. elegans), UNC84A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23353 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O94901 | |
SUN1 | |
SUN1 Fusion Protein Ag21117 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |