Läs mer
Sunitinib N-Oxide, TRC
Produktkod. 30010618
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010618 10 mg
30009507 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010618 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS82000510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 356068-99-0
Molekylformel C22 H27 F N4 O3
Molekylvikt (g/mol) 414.47
Synonym 1H-Pyrrole-3-carboxamide, N-[2-(diethyloxidoamino)ethyl]-5-[(Z)-(5-fluoro-1,2-dihydro-2-oxo-3H-indol-3-ylidene)methyl]-2,4-dimethyl-, 5-(5-Fluoro-2-oxo-1,2-dihydroindol-(3Z)-ylidenemethyl)-2,4-dimethyl-1H-pyrrole-3-carboxylic acid (2-diethyl-N-oxoaminoethyl)amide
IUPAC-namn N,N-diethyl-2-[[5-[(Z)-(5-fluoro-2-oxo-1H-indol-3-ylidene)methyl]-2,4-dimethyl-1H-pyrrole-3-carbonyl]amino]ethanamine oxide
LEDER CC[N+]([O-])(CC)CCNC(=O)c1c(C)[nH]c(\C=C\2/C(=O)Nc3ccc(F)cc23)c1C
Specifikationer
Kemiskt namn eller material Sunitinib N-Oxide
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C22H27FN4O3/c1-5-27(30,6-2)10-9-24-22(29)20-13(3)19(25-14(20)4)12-17-16-11-15(23)7-8-18(16)26-21(17)28/h7-8,11-12,25H,5-6,9-10H2,1-4H3,(H,24,29)(H,26,28)/b17-12-
Formel vikt 414.2067
Procent renhet >95%