missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUOX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15075-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Sulfite oxidase is a homodimeric protein localized to the intermembrane space of mitochondria. Each subunit contains a heme domain and a molybdopterin-binding domain. The enzyme catalyzes the oxidation of sulfite to sulfate, the final reaction in the oxidative degradation of the sulfur amino acids cysteine and methionine. Sulfite oxidase deficiency results in neurological abnormalities which are often fatal at an early age. Alternative splicing results in multiple transcript variants encoding identical proteins.Specifications
SUOX | |
Polyclonal | |
Unconjugated | |
SUOX | |
sulfite oxidase, Sulfite oxidase, mitochondrial, SUOX | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
211389, 6821, 81805 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry, Immunoprecipitation | |
0.75 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P51687, Q07116, Q8R086 | |
Suox | |
SUOX Fusion Protein Ag7146 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |