Learn More
SUOX Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558295
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RDGETVESGE VTIKGYAWSG GGRAVIRVDV SLDGGLTWQV AKLDGEEQRP RKAWAWRLWQ LKAPVPAGQK ELNIVCKAVD DGYNVQPDTV APIWNLRGVL SNAWHRVHVY V Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
Sulfite oxidase is a homodimeric protein localized to the intermembrane space of mitochondria. Each subunit contains a heme domain and a molybdopterin-binding domain. The enzyme catalyzes the oxidation of sulfite to sulfate, the final reaction in the oxidative degradation of the sulfur amino acids cysteine and methionine. Sulfite oxidase deficiency results in neurological abnormalities which are often fatal at an early age. Alternative splicing results in multiple transcript variants encoding identical proteins.Specifications
SUOX | |
Polyclonal | |
Unconjugated | |
SUOX | |
SO; sulfite oxidase; sulfite oxidase, mitochondrial; Suox | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6821 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P51687 | |
SUOX | |
Recombinant protein corresponding to Human SUOX | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only