Learn More
Cellpath Supa Mega Tissue Processing Cassettes
Product Code. 30226233
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 100
Product Code. Color Unit Size
30226233 White Pack of 100
1 options
Product Code. 30226233 Supplier Cellpath Supplier No. EAG010202A
  • ONLINE EXCLUSIVE PRICE
    1847.38 SEK
    1022.90 SEK / Paket med 100
    Save 824.48 SEK
    44% Av
 Restricted Item
Beräknad leverans: 01-03-2027
Widely adopted in research and toxicology laboratories. CellPath Supa Mega Tissue Processing Cassettes are ideal for embedding and processing larger tissue samples of prostrate, brain, breast and eye.
  • Plastic hinged lid
  • Rigid, durable
  • Excellent writing surfaces
  • Internal chamber is three times depth of the Supa Mega Slim and Supa Mega Mother Ship
  • Available in a range of colors
Specifications
Quantity 100/Pk.
Color White